2-Dodecylphenol ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
TCIAD2302-10GEA
0
CNY
TCIAD2302-10G
TCIAD2302-25G
2-Dodecylphenol ≥98.0% (通过GC)
2-Dodecylphenol
分子式:
C₁₈H₃₀O |
MDL编号:
MFCD00020106 CAS号: 5284-29-7 |
产品技术规格
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...