3-Undecylthiophene ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAU0056-5GEA
0
CNY
TCIAU0056-5G
TCIAU0056-1G
3-Undecylthiophene ≥98.0% (通过GC)
3-Undecylthiophene
分子式:
C₁₅H₂₆S 分子量: 238,4300 g/mol 沸点: 166°C |
MDL编号:
MFCD00130144 CAS号: 129607-86-9 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...