亚苄基酞 ≥99.0% (通过GC)

供应商: TCI EUROPE

Synonyms: 3-亚苄基酞

TCIAB1214-25GEA 0 CNY
TCIAB1214-25G
亚苄基酞 ≥99.0% (通过GC)
亚苄基酞
分子式: C₁₅H₁₀O₂
MDL编号: MFCD00005929
CAS号: 575-61-1
EINECS: 209-388-8
https://cn.vwr.com/stibo/web/std.lang.all/30/50/28933050.jpg

现在订购

产品技术规格

Purity (GC) min. 99.0 %
Melting point 100.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR