3-甲基-2-吡啶醛 ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
3-甲基吡啶-2-甲醛
TCIAM1470-1GEA
0
CNY
TCIAM1470-1G
3-甲基-2-吡啶醛 ≥98.0% (通过GC)
3-甲基-2-吡啶醛
分子式:
C₇H₇NO 分子量: 121,14 g/mol 沸点: 84°C 溶点: 8°C 闪点: 73°C |
MDL编号:
MFCD02181144 CAS号: 55589-47-4 |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...