4-Bromo-4'-pentylbiphenyl ≥99.0% (通过GC)
供应商: TCI EUROPE
TCIAB1305-5GEA
0
CNY
TCIAB1305-5G
4-Bromo-4'-pentylbiphenyl ≥99.0% (通过GC)
4-Bromo-4'-pentylbiphenyl
分子式:
C₁₇H₁₉Br 分子量: 303,2400 g/mol |
MDL编号:
MFCD00060106 CAS号: 63619-59-0 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 96.0 to 99.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...