Trimellitic acid tributyl ester ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAB0880-25MLEA 0 CNY
TCIAB0880-25ML
Trimellitic acid tributyl ester ≥98.0% (通过GC)
Trimellitic acid tributyl ester
分子式: C₂₁H₃₀O₆
沸点: 220°C
闪点: 215°C
MDL编号: MFCD00059424
CAS号: 1726-23-4
EINECS: 217-038-0
https://cn.vwr.com/stibo/web/std.lang.all/27/72/28932772.jpg

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR