96-well assay plates, round well, Axygen®
供应商: Corning Life Sciences
Corning® Axygen®
AXYGP-96-450R-C-SEA 423 CNY
AXYGP-96-450R-C-S AXYGP-96-450R-C
96-well assay plates, round well, Axygen®
微孔板 高温超导微孔板
Axygen® round well assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.
  • SBS footprint
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • Certified free of RNase, DNase and pyrogens