PCR plates, 384-well, Axygen®
供应商: Corning
AXYGPCR-384M2-C_UEA 0 CNY
AXYGPCR-384M2-C_U AXYGPCR-384-C_U
PCR plates, 384-well, Axygen®
PP plates, silicone mats. Designed to fit 384-well thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified RNase, DNase and endotoxin-safe for highest sample integrity
Designed to fit 384-well thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified RNase, DNase and endotoxin-safe for highest sample integrity