Sealing mat for 2,2 ml deep well blocks
供应商: Hamilton
HAMI6474-01EA 2393.28 CNY
HAMI6474-01
Sealing mat for 2,2 ml deep well blocks
Designed for sealing 2,2 ml deep 96-well blocks. DNase- and RNase-free, pyrogen-free (endotoxin-free), PCR inhibitor-free.