Real-Time PCR plates, 384-well, Eppendorf twin.tec® 384
供应商: Eppendorf
EPPE0030132.734EA 3225 CNY
EPPE0030132.734 EPPE0030132.750 EPPE0030132.742
Real-Time PCR plates, 384-well, Eppendorf twin.tec® 384
PC frame, white PP wells. The limiting factor in low volume Real-Time PCR is often the remaining intensity of fluorescence. The white wells of the Eppendorf twin.tec® real-time PCR plates reflect fluorescence much better than clear or frosted wells. Thus, lower levels of fluorescence are still detectable with the same instrument. Additionally, white wells significantly reduce interfering background fluorescence and lead to increased homogeneity of replicates and reproducible results. The rigidity of the polycarbonate frame ensure easy and reliable handling. The polypropylene wells guarantee excellent and fast temperature transfer to the sample.
  • White wells for better reflection
  • High mechanical stability
  • Effective sealing by raised well rims
The limiting factor in low volume Real-Time PCR is often the remaining intensity of fluorescence. The white wells of the Eppendorf twin.tec real-time PCR Plates reflect fluorescence much better than clear or frosted wells. Thus, lower levels of fluorescence are still detectable with the same instrument. Additionally, white wells significantly reduce interfering background fluorescence and lead to increased homogeneity of replicates and reproducible results. The rigidity of the polycarbonate frame ensure easy and reliable handling. The polypropylene wells guarantee excellent and fast temperature transfer to the sample.
  • White wells for better reflection
  • High mechanical stability
  • Effective sealing by raised well rims