2-Phenyl-2-(p-toluenesulphonyloxy)acetophenone ≥97.0% (通过HPLC)
供应商: TCI EUROPE
TCIAP1410-1GEA
0
CNY
TCIAP1410-1G
2-Phenyl-2-(p-toluenesulphonyloxy)acetophenone ≥97.0% (通过HPLC)
2-Phenyl-2-(p-toluenesulphonyloxy)acetophenone
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 105.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...