1-Allylimidazole ≥97.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Danger
|
TCIAA2207-25GEA
0
CNY
TCIAA2207-25G
1-Allylimidazole ≥97.0% (通过GC,滴定分析)
1-Allylimidazole
分子式:
C₆H₈N₂ 分子量: 108,14 g/mol 沸点: 116°C 闪点: 101°C |
MDL编号:
MFCD00037894 CAS号: 31410-01-2 EINECS: 250-618-1 UN号: 3267 ADR: 8,II |
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...