trans-4-(4-Ethylcyclohexyl)benzonitrile ≥98.0% (通过GC)
供应商: TCI EUROPE
|
Danger
|
TCIAE0703-5GEA
0
CNY
TCIAE0703-5G
trans-4-(4-Ethylcyclohexyl)benzonitrile ≥98.0% (通过GC)
trans-4-(4-Ethylcyclohexyl)benzonitrile
分子式:
C₁₅H₁₉N 分子量: 213,32 g/mol |
MDL编号:
MFCD09750932 CAS号: 72928-54-2 EINECS: 277-084-2 UN号: 3439 ADR: 6.1,III |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...