1-苯基双胍 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI EUROPE

TCIAP1002-10GEA 0 CNY
TCIAP1002-10G TCIAP1002-5G
1-苯基双胍 ≥98.0% (通过HPLC,滴定分析)
1-苯基双胍
分子式: C₈H₁₁N₅
分子量: 177,2064 g/mol
MDL编号: MFCD00179077
CAS号: 102-02-3
EINECS: 202-998-5
https://cn.vwr.com/stibo/web/std.lang.all/16/37/28991637.jpg

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR