2-Furfurylthio-3-methylpyrazine ≥95.0% (通过GC)
供应商: TCI EUROPE
TCIAF0222-5GEA
0
CNY
TCIAF0222-5G
2-Furfurylthio-3-methylpyrazine ≥95.0% (通过GC)
2-Furfurylthio-3-methylpyrazine
分子式:
C₁₀H₁₀N₂OS |
MDL编号:
MFCD00055031 CAS号: 59303-07-0 EINECS: 261-690-9 |
产品技术规格
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...