(R)-(-)-3-Aminopyrrolidine dihydrochloride ≥98.0% (通过滴定分析)

Warning

TCIAA1053-5GEA 1475 CNY
TCIAA1053-5G
(R)-(-)-3-Aminopyrrolidine dihydrochloride ≥98.0% (通过滴定分析)
(R)-(-)-3-Aminopyrrolidine dihydrochloride
分子式: C₄H₁₂Cl₂N₂
分子量: 159,06 g/mol
MDL编号: MFCD00070604
CAS号: 116183-81-4
https://cn.vwr.com/stibo/web/std.lang.all/88/01/28928801.jpg

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to lump
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -0.9 to -1.3 deg (C=10, H2O)
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR