1-Boc-2-methylpiperazine

供应商: LOCAL DEALS CHINA

CHLCR03097425141EA 0 CNY
CHLCR03097425141
1-Boc-2-methylpiperazine
1-Boc-2-methylpiperazine
分子式: C₁₀H₂₀N₂O₂
分子量: 200,2780 g/mol
MDL编号: MFCD02179168
CAS号: 120737-78-2

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR