胡椒基胺 ≥97.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Danger
|
Synonyms:
3,4-亚甲二氧基苯甲胺
, 3,4-亚甲二氧基苄胺
TCIAP1057-25GEA
0
CNY
TCIAP1057-25G
胡椒基胺 ≥97.0% (通过GC,滴定分析)
胡椒基胺
分子式:
C₈H₉NO₂ 分子量: 151,17 g/mol 沸点: 132°C |
MDL编号:
MFCD00005840 CAS号: 2620-50-0 EINECS: 220-056-1 UN号: 2735 ADR: 8,III |
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...