二甲硝咪唑 ≥98.0% (通过GC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
Synonyms:
1,2-二甲基-5-硝基咪唑
TCIAD1564-25GEA
0
CNY
TCIAD1564-25G
二甲硝咪唑 ≥98.0% (通过GC,滴定分析)
二甲硝咪唑
分子式:
C₅H₇N₃O₂ 分子量: 141,13 g/mol 存储条件: Refrigerator |
MDL编号:
MFCD00047046 CAS号: 551-92-8 EINECS: 209-001-2 |
产品技术规格
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 137.0 to 141.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...