4-溴重氮苯四氟化硼酸盐 ≥97.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Danger
|
TCIAB3114-5GEA
0
CNY
TCIAB3114-5G
4-溴重氮苯四氟化硼酸盐 ≥97.0% (通过HPLC,滴定分析)
4-溴重氮苯四氟化硼酸盐
分子式:
C₆H₄BBrF₄N₂ 分子量: 270,83 g/mol 溶点: 133°C |
MDL编号:
MFCD00011894 CAS号: 673-40-5 EINECS: 211-608-2 UN号: 1759 ADR: 8,III |
产品技术规格
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...