5-Chlorosalicylamide ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAC0787-5GEA
0
CNY
TCIAC0787-5G
TCIAC0787-25G
5-Chlorosalicylamide ≥98.0% (通过HPLC,滴定分析)
5-Chlorosalicylamide
分子式:
C₇H₆ClNO₂ 分子量: 171,58 g/mol |
MDL编号:
MFCD00007980 CAS号: 7120-43-6 |
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 233.0 to 237.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...