(±)-Z-α-膦酰甘氨酸三甲酯 95%
供应商: Thermo Scientific
|
Warning
|
Synonyms:
(±)-三甲基 Z-α-膦酰甘氨酸酯
, (±)-甲基 2-苄氧羰基氨基-2-(二甲氧基氧膦基)乙酸酯
ACRO370040050EA
0
CNY
ACRO370040050
(±)-Z-α-膦酰甘氨酸三甲酯 95%
(±)-Z-α-膦酰甘氨酸三甲酯
分子式:
C₁₃H₁₈NO₇P 分子量: 331,27 溶点: 77…80 °C 存储条件: Ambient |
MDL编号:
MFCD00043304 CAS号: 88568-95-0 |
产品技术规格
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 77 to 80 °C |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...