1-(6-Methoxy-2-naphthyl)ethanol ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAH1270-5GEA
0
CNY
TCIAH1270-5G
1-(6-Methoxy-2-naphthyl)ethanol ≥98.0% (通过GC)
1-(6-Methoxy-2-naphthyl)ethanol
分子式:
C₁₃H₁₄O₂ 分子量: 202,25 g/mol |
MDL编号:
MFCD01632581 CAS号: 77301-42-9 |
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...