N,N-Dimethyl-4-(2-pyridylazo)aniline ≥98.0% (通过HPLC,滴定分析)
供应商: TCI EUROPE
|
Warning
|
TCIAP0617-1GEA
0
CNY
TCIAP0617-1G
N,N-Dimethyl-4-(2-pyridylazo)aniline ≥98.0% (通过HPLC,滴定分析)
N,N-Dimethyl-4-(2-pyridylazo)aniline
分子式:
C₁₃H₁₄N₄ 分子量: 226,28 g/mol |
MDL编号:
MFCD00010435 CAS号: 13103-75-8 EINECS: 236-026-6 |
产品技术规格
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 110.0 to 114.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...