2,5-Bis(4-(diethylamino)phenyl)-1,3,4-oxadiazole ≥98.0% (通过滴定分析)

供应商: TCI EUROPE
Warning

TCIAB1066-25GEA 0 CNY
TCIAB1066-25G
2,5-Bis(4-(diethylamino)phenyl)-1,3,4-oxadiazole ≥98.0% (通过滴定分析)
2,5-Bis(4-(diethylamino)phenyl)-1,3,4-oxadiazole
分子式: C₂₂H₂₈N₄O
分子量: 364,49 g/mol
MDL编号: MFCD00010070
CAS号: 1679-98-7
EINECS: 216-851-8
https://cn.vwr.com/stibo/web/std.lang.all/29/22/28932922.jpg

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR