1-Isopropylimidazole ≥97.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Warning

TCIAI0845-25GEA 345 CNY
TCIAI0845-25G
1-Isopropylimidazole ≥97.0% (通过GC,滴定分析)
1-Isopropylimidazole
分子式: C₆H₁₀N₂
分子量: 110,1600 g/mol
沸点: 104°C
MDL编号: MFCD00234239
CAS号: 4532-96-1
https://cn.vwr.com/stibo/web/std.lang.all/27/32/28972732.jpg

现在订购

产品技术规格

Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR