2,2'-Diiodobiphenyl ≥98.0% (通过GC)

供应商: TCI EUROPE

TCIAD2745-1GEA 0 CNY
TCIAD2745-1G
2,2'-Diiodobiphenyl ≥98.0% (通过GC)
2,2'-Diiodobiphenyl
分子式: C₁₂H₈I₂
分子量: 406,0009 g/mol
MDL编号: MFCD00017622
CAS号: 2236-52-4
UN号: 3152
ADR: 9,II
https://cn.vwr.com/stibo/web/std.lang.all/98/71/28979871.jpg

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR