对苯二甲胺 ≥99.0% (通过GC,滴定分析)

供应商: TCI EUROPE
Danger

Synonyms: α,α′-二氨基对二甲苯1,4-苯二甲胺

TCIAD1018-25GEA 720 CNY
TCIAD1018-25G TCIAD1018-500G TCIAD1018-100G
对苯二甲胺 ≥99.0% (通过GC,滴定分析)
对苯二甲胺
分子式: C₈H₁₂N₂
分子量: 136,20 g/mol
MDL编号: MFCD00009821
CAS号: 539-48-0
EINECS: 208-719-3
UN号: 3259
ADR: 8,III
https://cn.vwr.com/stibo/web/std.lang.all/62/16/28976216.jpg

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 60.0 to 64.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR