二苯基乙酸甲酯 ≥98.0% (通过GC)
供应商: TCI EUROPE
TCIAD2888-25GEA
0
CNY
TCIAD2888-25G
二苯基乙酸甲酯 ≥98.0% (通过GC)
二苯基乙酸甲酯
分子式:
C₁₅H₁₄O₂ 分子量: 226,28 g/mol |
MDL编号:
MFCD00025869 CAS号: 3469-00-9 EINECS: 222-431-5 |
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...