4-(三甲基硅)-3-丁炔-2-醇 ≥97.0% (通过GC)

供应商: TCI EUROPE
Warning

TCIAT1926-5GEA 550 CNY
TCIAT1926-5G
4-(三甲基硅)-3-丁炔-2-醇 ≥97.0% (通过GC)
4-(三甲基硅)-3-丁炔-2-醇
分子式: C₇H₁₄OSi
分子量: 142,27
闪点: 32°C
存储条件: Refrigerator
MDL编号: MFCD00190213
CAS号: 6999-19-5
UN号: 1993
ADR: 3,III
https://cn.vwr.com/stibo/web/std.lang.all/58/81/28995881.jpg

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR