1-(4-Methylphenyl)cyclopropanecarboxylic acid 98%
供应商: Thermo Scientific
|
Warning
|
ACRO170700100EA
0
CNY
ACRO170700100
1-(4-Methylphenyl)cyclopropanecarboxylic acid 98%
1-(4-Methylphenyl)cyclopropanecarboxylic acid
分子式:
C₁₁H₁₂O₂ 分子量: 176,2100 g/mol 溶点: 109…114 °C 存储条件: Ambient |
MDL编号:
MFCD00066918 CAS号: 83846-66-6 EINECS: 281-043-4 |
产品技术规格
Appearance | Beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 109 to 114 °C |
Titration with NaOH | ≥97.5 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...