Tubing holder for beakers, SP Bel-Art
供应商: Bel-Art Products, a part of SP
BELAF182350000EA 0 CNY
BELAF182350000
Tubing holder for beakers, SP Bel-Art
夹具 油管夹
This PP clip holds flexible tubing steady and fits securely onto the edge of beakers or pitchers up to ¼" thick.
  • 5,1 cm (2") long arm has a hole to accept tubing up to 9,5 mm (⅜") Ext. Ø
  • Autoclavable