The anion exchange HPLC column is suitable for isocratic determinations of monosaccharides, disaccharides and linear polysaccharides in a variety of matrices. This specialised column combined with pulsed amperometric detection (PAD), delivers high-resolution separations. Approved for use in a variety of official methods for the analysis of foods.
- Suited for the analysis of foods for nutritional labelling
- Ideal for the separation of neutral and acidic monosaccharides
- Lower sensitivity than the Dionex™ CarboPac™ PA10 or PA20 columns
The unique Dionex™ MicroBead™ pellicular structure of the Dionex™ CarboPac™ PA1 resin gives it stability from pH 0 to 14 at all concentrations of buffer salts. The pellicular resin structure provides mass transfer resulting in fast gradient re-equilibration.