XT starter is a low-profile open platform cooler that keeps samples at a uniform 2 °C for over for hours without ice, batteries or electricity. XT starter accommodates most CoolRack and CoolSink sample modules, making it a versatile and flexible tool for a variety of applications. The low profile and small footprint make it ideal for use in the hood. The built-in 1 - 8 °C temperature indicator provides visual assurance of operational temperature while working.
XT starter includes one XT holder base and one XT cooling core. To extend the cooling duration, keep an additional XT cooling core in the freezer and replace the core as needed.
To assemble, simply pre-freeze the XT cooling core in a -20 °C cooler until frozen. When ready to use, rest the XT cooling core on the bench until the temperature indicator registers 1 °C (approx. 10 min). Then insert the XT cooling core into the XT holder and place a CoolRack or CoolSink sample module on top.
订购须知 For replacement cooling cores see VWR catalog number 216-0963.