Sample bags, Bitran® series
供应商: Com-Pac
Bitran®
CPAC4753-PEEA 1215 CNY
CPAC4753-PE CPAC4752-PE CPAC4754-PE CPAC4755-PE
Sample bags, Bitran® series
样品袋
These bags are ideal for for short-term handling and storage of specimens and are manufactured from durable 2,8 mil PE that remains flexible when frozen.
  • Patented manufacturing process eliminates leaks and offers unprecedented safety
  • Double-track zipper is airtight and easy to close
  • Appropriate for use as a primary container for most specimens
  • Custom sizes and printing available
Packaging: Bags are packaged in packs of 250 each, four packs per case.