Handheld colorimeter, DR 900
供应商: Hach
HACH9385100EA 14975 CNY
HACH9385100
Handheld colorimeter, DR 900
比色计 多参数色度计
The DR 900 allows quick and easy access to your most-used testing methods. This colorimeter is waterproof, dustproof and field durable. With an intuitive user interface, easy data transfer abilities, and the ability to test up to 90 of the most common water testing methods, the DR 900 makes water analysis in harsh field environments a little less challenging.
  • Field ready in every way possible
  • Intuitive user interface
  • Simple data communication
  • IP 67 enclosure rating
Delivery information: DR 900 instrument supplied with USB cable with mini-USB connector, 4×AA alkaline batteries, 2× glass sample cells Ø 1 inch (25 mm) with graduations at 10, 20, 25 ml, 2× sample cells 1 cm/10 ml, sample cell adapter, instrument procedures CD, and multilingual user manual.