Beta blocking apron, SP Bel-Art
供应商: Bel-Art Products, a part of SP
BELAF246020000EA 5350 CNY
BELAF246020000
Beta blocking apron, SP Bel-Art
This lightweight, yet effective Beta-blocking neoprene apron blocks up to 96% of the high energy beta rays from Strontium-90 and over 98% of the beta rays from Phosphorous-32. Perfect for safeguarding against chemical splashes.
  • Strap system places weight of apron on the shoulders instead of the neck
  • Intended for use in conjunction with radiation protection shields
Weight: 1,5 kg