Cut resistant gloves, Whizard® Silver Talon®
供应商: Wells Lamont
Whizard®
WELL134527_UEA 0 CNY
WELL134527_U
Cut resistant gloves, Whizard® Silver Talon®
These gloves feature are manufactured from a yarn composition of high performance fibers and stainless steel which provides exceptional protection against cuts and slashes.
  • 'Second Skin' fit and flexibility
  • Unmatched combination of cut resistance, dexterity and durability
  • Unique coloured cuff is highly visible
  • Ambidextrous
Ideal for use in the meat processing industry.
Certifications: ANSI Level 5
Food contact approved