Vertical electrophoresis system, Criterion™
供应商: Bio-Rad
BRAD165-6001EA 0 CNY
BRAD165-6001
Vertical electrophoresis system, Criterion™
电泳系统 垂直电泳系统
Designed to run one or two vertical, midi-format Criterion precast gels. This cell includes a buffer tank, lid with power cables, and three sample loading guides.