Bottle-top dispensers, SofTop™
供应商: Hamilton
HAMI51249-01EA 4999.41 CNY
HAMI51249-01 HAMI51249-60 HAMI51249-30
Bottle-top dispensers, SofTop™
Fixed or variable volume, autoclavable.
  • Micrometric adjustment knob for fine dispensing
  • Ceramic piston made of a high purity for the best chemical and thermal inertia
  • Certificate of conformity and quality
The SofTop Quik dispenser is safe and has a screw-fitted suction hose, eliminating the possibility of air bubbles during aspiration.