Immobiline™ drystrip cover fluid
供应商: Cytiva
GEHE17-1335-01EA 0 CNY
GEHE17-1335-01
Immobiline™ drystrip cover fluid
凝胶电泳 预制凝胶
This purified fraction of mineral oil protects immobiline drystrip gels from drying and from the atmosphere during the first dimension of 2-D electrophoresis
  • Boiling point at 330°C
  • Insoluble in cold and hot water
  • Storage conditions from 18 to 25 °C
The fluid is suitable as a contact between cooling plate and gel backing film in horizontal electrophoresis.