Connectors, tubing, 28-piece fitting kit, SP Bel-Art
供应商: Bel-Art
SCIENCEWARE®
BELAF196260000EA 1369.44 CNY
BELAF196260000
Connectors, tubing, 28-piece fitting kit, SP Bel-Art
连接器 管接头
Most common PP or PE fittings, translucent, for the lab.
  • PP fittings are autoclavable and FDA conform
  • PE quick disconnect fittings are not FDA grade or autoclavable
  • Compatible with tubing from 3,2 to 12,7 mm
  • Inluces PS organiser box with additional room for other fittings and parts
订购须知 The kit contains 6 straight tubing connectors, 7 stepped tubing connectors, 5 Wye (Y) tubing connectors, 3 'T' shaped tubing connectors, 5 quick disconnects and 2 flow control plastic tubing clamps.