PCR plates, 96-well, for MegaBACE™ systems, Axygen®
供应商: Corning Life Sciences
AXYGPCR-96-MB-VEA 0 CNY
AXYGPCR-96-MB-V AXYGPCR-96-MB-A AXYGPCR-96MB-W
PCR plates, 96-well, for MegaBACE™ systems, Axygen®
Virgin PP. These plates feature elevated chimney wells that are slightly taller and narrower than typical chimney wells, making these plates ideal for use with Amersham™ MegaBACE™ DNA analysis systems.
  • Ultra-thin walls accelerate heat transfer to provide shorter cycle times
  • Certified free from DNase, RNase and pyrogens
  • Working volume: 300 µl
Optional AxyMat™ silicone sealing mat is autoclavable and freezable. Mats are pierceable and self-sealing to prevent escape of evaporated aerosols and pathogens during normal use or storage. Alphanumeric grid system corresponds to 96-well plates.