These flock-lined gloves made of a high performance nitrile compound provide an outstanding combination of chemical resistance and mechanical strength.
- Offers superior snag, puncture and abrasion protection over rubber or neoprene gloves
- Suitable for heavy-duty cleaning applications
- High levels of flexibility, comfort and dexterity
- Won't swell, weaken or degrade, and does not promote contact dermatitis
- Straight cuff, sandpatch finish
Suitable for heavy-duty cleaning applications, food processing, janitorial and maintenance work.
Certifications: EN 388 (3101)
Food contact approved