Bulb pipettes, BLAUBRAND®, two marks, class AS, USP
供应商: Brand
BRNDUSP29721EA 0 CNY
BRNDUSP29721 BRNDUSP29725
Bulb pipettes, BLAUBRAND®, two marks, class AS, USP
AR-Glas®
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Waiting time 5 seconds
  • With DE-M marking
Certifications: DIN EN ISO 648; USP certificate