Volumetric pipettes, class A, color-coded, graduated, serialised/certified, PYREX®
供应商: Corning
PYREX®
CORN7101-20EA 0 CNY
CORN7101-10 CORN7101-20 CORN7101-100 CORN7101-15 CORN7101-25 CORN7101-1 CORN7101-2 CORN7101-3 CORN7101-4 CORN7101-5 CORN7101-50
Volumetric pipettes, class A, color-coded, graduated, serialised/certified, PYREX®
移液管和移液器 刻度移液管
Pipettes are calibrated to Class A tolerances in accordance with ASTM E-542 and ASTM E-969.
  • Sizes 1 ml through 25 ml are color-coded for ease in sorting and selection
  • Coloured graduations are enamelled on to the glass
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Delivery information: Each pipette is individually serialised and supplied with a certificate of identity and capacity, traceable to NIST standards.