Volumetric flasks, class A, glass, clear, glass stopper
供应商: Corning Life Sciences
Corning®
CORN5640-4LEA 0 CNY
CORN5640-10FO CORN5640-5L CORN5640-200FO CORN5640-25FO CORN5640-10 CORN5640-25 CORN5640-5 CORN5640-4L CORN5640-1 CORN5640-500FO CORN5640-250 CORN5640-3L CORN5640-100 CORN5640-50FO CORN5640-500 CORN5640-100FO CORN5640-250FO CORN5640-1LFO CORN5640-200 CORN5640-2 CORN5640-1L CORN5640-2L CORN5640-6L CORN5640-50
Volumetric flasks, class A, glass, clear, glass stopper
烧瓶 容量瓶
PYREX® borosilicate glass, class A, clear, with glass stopper.
  • Strength increased through machine-blown bodies
  • Sharp graduation line and permanent and large white block letters make the inscription easy to read
ASTM E-694, E-542, E-288
Certifications: ASTM E-694, E-542, E-288