Culture tubes with phenolic screw caps
供应商: Corning Life Sciences
PYREX® Corning®
CORN9825-16XXEA 0 CNY
CORN9826-25X CORN9825-22 CORN9826-25 CORN9825-16XX CORN9826-16X CORN9825-16XX_U1 CORN9825-25 CORN9826-20X CORN9825-16X_U1 CORN9825-16 CORN9825-25X CORN9826-13 CORN9825-16X CORN9826-16XX CORN9826-16 CORN9825-13 CORN9825-20X CORN9826-16_U CORN9825-38 CORN9826-20 CORN9825-20
Culture tubes with phenolic screw caps
管子 培养管
PYREX®, borosilicate glass, round bottom with screw cap.
  • Excellent chemical resistance
  • Excellent temperature resistance
Phenolic caps are resistant to temperature and steam in autoclaving. White enamel marking spot allows excellent surface for pencil notations. Pencil marks are easily erased. Phenolic cap with PTFE liner firmly bonded to the glued-in rubber liner by providing a sure and highly inert sealing face.