Separating funnels, Squibb
供应商: Corning Life Sciences
PYREX®
CORN6402-2LEA 0 CNY
CORN6402-60FO CORN6402-500 CORN6402-1L CORN6402-250FO CORN6402-4L CORN6402-60 CORN6402-2L CORN6402-250 CORN6402-125 CORN6402-6L
Separating funnels, Squibb
漏斗 分液漏斗
PYREX®, borosilicate glass, with glass stopper.
With socket and plain stem. Available with interchangeable PTFE key for grease-free performance.