Reagent bottles, narrow neck, with standard taper stoppers, PYREX®
供应商: Corning
PYREX®
CORN1500-250EA 0 CNY
CORN1500-2L CORN1500-500 CORN1500-1L CORN1500-250
Reagent bottles, narrow neck, with standard taper stoppers, PYREX®
玻璃瓶
PYREX® borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These narrow mouth reagent storage bottles have sturdy ground necks with standard taper stoppers. Replacement stoppers are available.